siRNA Details
Gene Symbol: Rragb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004154.2 | NP_001004154.1 | 3, 4 | 756 | ||
GenBank mRNA | BC069180.1 | - | 3, 4 | 714 | 6810543 | |
BC070407.1 | - | 3, 4 | 733 | 6808979 |
