siRNA Details
| Gene Symbol: Fat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029988.2 | NP_001025159.1 | 14 | 10011 | ||
| GenBank mRNA | AK173031.1 | - | 6 | 1909 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fat2| Gene Symbol: Fat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029988.2 | NP_001025159.1 | 14 | 10011 | ||
| GenBank mRNA | AK173031.1 | - | 6 | 1909 | ||