siRNA Details
| Gene Symbol: Ift52 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172150.4 | NP_742162.2 | 10 | 943 | ||
| GenBank mRNA | AK088467.1 | - | 10 | 944 | ||
| AK144188.1 | - | 10 | 1043 | |||
| AK167194.1 | - | 10 | 959 | |||
| AK178241.1 | - | 4 | 310 | |||
| BC037708.1 | - | 10 | 912 | 4222723 | ||

Ift52
Genomic Map