siRNA Details
| Gene Symbol: Rbm47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127382.2 | NP_001120854.1 | 5 | 606 | ||
| NM_001291226.1 | NP_001278155.1 | 4 | 806 | |||
| NM_139065.3 | NP_620704.1 | 4 | 364 | |||
| NM_178446.4 | NP_848541.2 | 4 | 806 | |||
| GenBank mRNA | AK035620.1 | - | 4 | 757 | ||
| AK085733.1 | - | 4 | 798 | |||
| AK089362.1 | - | 4 | 729 | |||
| AK142584.1 | - | 4 | 784 | |||
| AK170082.1 | - | 5 | 561 | |||
| BC013481.1 | - | 4 | 376 | 4240441 | ||
| BC034195.1 | - | 3 | 312 | 4984936 | ||
| BC112902.1 | - | 4 | 806 | 4978873 | ||

Rbm47
Genomic Map