siRNA Details
| Gene Symbol: Otoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139310.1 | NP_647471.1 | 24 | 2720 | ||
| GenBank mRNA | AK157904.1 | - | 24 | 2752 | ||
| AK158280.1 | - | 14 | 1693 | |||
| AK158326.1 | - | 14 | 1689 | |||
| AY055122.1 | - | 24 | 2720 | |||

Otoa
Genomic Map