siRNA Details
| Gene Symbol: Zfp277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172575.3 | NP_766163.2 | 4, 5 | 798 | ||
| NM_178845.3 | NP_849173.2 | 3, 4 | 420 | |||
| GenBank mRNA | AK075945.1 | - | 3, 4 | 411 | ||
| AK077401.1 | - | 4, 5 | 783 | |||
| AK145060.1 | - | 3, 4 | 416 | |||
| AK156529.1 | - | 3, 4 | 401 | |||
| BC043453.1 | - | 3, 4 | 411 | 5004865 | ||

Zfp277
Genomic Map