siRNA Details
Gene Symbol: Emilin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145158.3 | NP_660140.1 | 8 | 3128 | ||
GenBank mRNA | AF468645.1 | - | 8 | 3079 | ||
AK154230.1 | - | 8 | 3098 | |||
AK155037.1 | - | 8 | 3109 | |||
AK155306.1 | - | 8 | 3122 | |||
AK155366.1 | - | 8 | 3109 | |||
AK155554.1 | - | 8 | 3124 | |||
AK155797.1 | - | 8 | 3096 | |||
AK155878.1 | - | 8 | 3127 | |||
AK155879.1 | - | 8 | 3123 | |||
AK155909.1 | - | 8 | 3128 | |||
AK155913.1 | - | 8 | 3098 | |||
AK155918.1 | - | 8 | 3156 | |||
AK170081.1 | - | 8 | 3122 | |||
AK170391.1 | - | 8 | 3098 | |||
AK170801.1 | - | 8 | 3129 | |||
AK170822.1 | - | 8 | 3150 | |||
AK170953.1 | - | 8 | 3144 | |||
AK171015.1 | - | 8 | 3150 | |||
AK171020.1 | - | 8 | 3116 | |||
AK171303.1 | - | 8 | 3100 | |||
AK171309.1 | - | 8 | 3125 | |||
AK171579.1 | - | 8 | 3099 | |||
AK171592.1 | - | 8 | 3098 | |||
AK171616.1 | - | 8 | 3099 | |||
BC053753.1 | - | 8 | 3412 | 6510220 | ||
GQ872457.1 | - | 8 | 3023 | |||
GQ872460.1 | - | 8 | 3023 |
