siRNA Details
Gene Symbol: Rhobtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153514.5 | NP_705734.4 | 8 | 2332 | ||
GenBank mRNA | AF420001.1 | - | 7 | 1823 | ||
AK038196.1 | - | 8 | 2424 | |||
AK081100.1 | - | 8 | 2261 | |||
BC026836.1 | - | 8 | 2313 | 4242008 | ||
BC056954.1 | - | 8 | 1937 | 5685456 |
