siRNA Details
Gene Symbol: Rhobtb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153514.5 | NP_705734.4 | 5 | 1768 | ||
GenBank mRNA | AF420001.1 | - | 4 | 1259 | ||
AK037927.1 | - | 4 | 2295 | |||
AK038196.1 | - | 5 | 1860 | |||
AK081100.1 | - | 5 | 1697 | |||
AK122354.1 | - | 5 | 1749 | |||
AK171295.1 | - | 11 | 2383 | |||
BC026836.1 | - | 5 | 1749 | 4242008 | ||
BC056954.1 | - | 5 | 1373 | 5685456 |
