siRNA Details
Gene Symbol: Trpv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145099.2 | NP_659567.2 | 8 | 880 | ||
GenBank mRNA | AF510316.1 | - | 8 | 880 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trpv3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145099.2 | NP_659567.2 | 8 | 880 | ||
GenBank mRNA | AF510316.1 | - | 8 | 880 |