siRNA Details
Gene Symbol: Obox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145710.2 | NP_663756.2 | 3 | 1085 | ||
GenBank mRNA | BC128024.1 | - | 2 | 1024 | 40136487 | |
BG069935.2 | - | 1 | 248 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Obox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145710.2 | NP_663756.2 | 3 | 1085 | ||
GenBank mRNA | BC128024.1 | - | 2 | 1024 | 40136487 | |
BG069935.2 | - | 1 | 248 |