siRNA Details
| Gene Symbol: Ropn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145852.2 | NP_665851.2 | 4 | 647 | ||
| GenBank mRNA | AF305427.1 | - | 4 | 604 | ||
| AK131675.1 | - | 4 | 696 | |||
| BC048680.1 | - | 4 | 583 | 6774441 | ||
| BC060990.1 | - | 4 | 591 | 6774094 | ||

Ropn1l
Genomic Map