siRNA Details
| Gene Symbol: Olfr479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011742.1 | NP_001011742.1 | 1 | 659 | ||
| GenBank mRNA | BC150837.1 | - | 1 | 686 | 9087748 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr479| Gene Symbol: Olfr479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011742.1 | NP_001011742.1 | 1 | 659 | ||
| GenBank mRNA | BC150837.1 | - | 1 | 686 | 9087748 | |
Set Membership: |
|