siRNA Details
Gene Symbol: Olfr426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206926.1 | NP_001193855.1 | 1 | 475 |
Gene Symbol: Olfr427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207158.1 | NP_997041.1 | 1 | 475 | ||
GenBank mRNA | BC104104.2 | - | 1 | 475 | 40043137 | |
BC106808.2 | - | 1 | 475 | 40042917 |
