siRNA Details
Gene Symbol: Olfr426 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206926.1 | NP_001193855.1 | 1 | 621 |
Gene Symbol: Olfr427 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207158.1 | NP_997041.1 | 1 | 621 | ||
GenBank mRNA | BC104103.2 | - | 1 | 621 | 40043136 | |
BC104104.2 | - | 1 | 621 | 40043137 | ||
BC106807.2 | - | 1 | 621 | 40042912 | ||
BC106808.2 | - | 1 | 621 | 40042917 |
