siRNA Details
| Gene Symbol: Olfr506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011871.1 | NP_001011871.1 | 1 | 445 | ||
| GenBank mRNA | BC134384.1 | - | 449 | |||
| BC147670.1 | - | 1 | 474 | 9056643 | ||
| BC147675.1 | - | 1 | 474 | 9056648 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr506| Gene Symbol: Olfr506 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011871.1 | NP_001011871.1 | 1 | 445 | ||
| GenBank mRNA | BC134384.1 | - | 449 | |||
| BC147670.1 | - | 1 | 474 | 9056643 | ||
| BC147675.1 | - | 1 | 474 | 9056648 | ||