siRNA Details
| Gene Symbol: Olfr94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011518.1 | NP_001011518.1 | 1 | 40 | ||
| GenBank mRNA | BC139173.1 | - | 1 | 85 | 8862195 | |
| BC139177.1 | - | 1 | 85 | 8862199 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr94| Gene Symbol: Olfr94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011518.1 | NP_001011518.1 | 1 | 40 | ||
| GenBank mRNA | BC139173.1 | - | 1 | 85 | 8862195 | |
| BC139177.1 | - | 1 | 85 | 8862199 | ||