siRNA Details
| Gene Symbol: Olfr845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207145.2 | NP_997028.1 | 1 | 827 | ||
| GenBank mRNA | BC127982.1 | - | 1 | 828 | 40135240 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr845| Gene Symbol: Olfr845 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_207145.2 | NP_997028.1 | 1 | 827 | ||
| GenBank mRNA | BC127982.1 | - | 1 | 828 | 40135240 | |