siRNA Details
| Gene Symbol: Olfr1311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146274.1 | NP_666386.1 | 1 | 764 | ||
| GenBank mRNA | BC127967.1 | - | 1 | 768 | 40135022 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1311| Gene Symbol: Olfr1311 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146274.1 | NP_666386.1 | 1 | 764 | ||
| GenBank mRNA | BC127967.1 | - | 1 | 768 | 40135022 | |