siRNA Details
| Gene Symbol: Olfr125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146290.2 | NP_666402.2 | 1 | 887 | ||
| GenBank mRNA | BC116810.1 | - | 1 | 887 | 40126129 | |
| BC116812.1 | - | 1 | 887 | 40126131 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr125| Gene Symbol: Olfr125 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146290.2 | NP_666402.2 | 1 | 887 | ||
| GenBank mRNA | BC116810.1 | - | 1 | 887 | 40126129 | |
| BC116812.1 | - | 1 | 887 | 40126131 | ||