siRNA Details
Gene Symbol: Olfr1449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146303.1 | NP_666415.1 | 1 | 99 | ||
GenBank mRNA | BC133705.1 | - | 1 | 103 | 40135405 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146303.1 | NP_666415.1 | 1 | 99 | ||
GenBank mRNA | BC133705.1 | - | 1 | 103 | 40135405 |