siRNA Details
Gene Symbol: Olfr657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146312.2 | NP_666424.2 | 1 | 590 | ||
GenBank mRNA | BC104311.1 | - | 1 | 585 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146312.2 | NP_666424.2 | 1 | 590 | ||
GenBank mRNA | BC104311.1 | - | 1 | 585 |