siRNA Details
| Gene Symbol: Olfr657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146312.2 | NP_666424.2 | 1 | 545 | ||
| GenBank mRNA | BC104311.1 | - | 1 | 540 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr657| Gene Symbol: Olfr657 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146312.2 | NP_666424.2 | 1 | 545 | ||
| GenBank mRNA | BC104311.1 | - | 1 | 540 | ||