siRNA Details
| Gene Symbol: Olfr1495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146344.1 | NP_666456.1 | 1 | 521 | ||
| GenBank mRNA | BC125566.1 | - | 1 | 558 | 40130081 | |
| BC137879.1 | - | 1 | 556 | 8860898 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1495| Gene Symbol: Olfr1495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146344.1 | NP_666456.1 | 1 | 521 | ||
| GenBank mRNA | BC125566.1 | - | 1 | 558 | 40130081 | |
| BC137879.1 | - | 1 | 556 | 8860898 | ||