siRNA Details
| Gene Symbol: Olfr720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146392.1 | NP_666504.1 | 1 | 488 | ||
| GenBank mRNA | BC120861.1 | - | 1 | 525 | 40129784 | |
| BC132043.1 | - | 1 | 525 | 40130320 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr720| Gene Symbol: Olfr720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146392.1 | NP_666504.1 | 1 | 488 | ||
| GenBank mRNA | BC120861.1 | - | 1 | 525 | 40129784 | |
| BC132043.1 | - | 1 | 525 | 40130320 | ||