siRNA Details
| Gene Symbol: Olfr1279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146393.1 | NP_666505.1 | 1 | 198 | ||
| GenBank mRNA | BC113137.1 | - | 1 | 198 | 40042162 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1279| Gene Symbol: Olfr1279 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146393.1 | NP_666505.1 | 1 | 198 | ||
| GenBank mRNA | BC113137.1 | - | 1 | 198 | 40042162 | |