siRNA Details
Gene Symbol: Olfr1279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146393.1 | NP_666505.1 | 1 | 110 | ||
GenBank mRNA | BC113137.1 | - | 1 | 110 | 40042162 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1279 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146393.1 | NP_666505.1 | 1 | 110 | ||
GenBank mRNA | BC113137.1 | - | 1 | 110 | 40042162 |