siRNA Details
Gene Symbol: Olfr1420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146410.1 | NP_666522.1 | 1 | 600 | ||
GenBank mRNA | BC120832.1 | - | 1 | 624 | 40129755 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146410.1 | NP_666522.1 | 1 | 600 | ||
GenBank mRNA | BC120832.1 | - | 1 | 624 | 40129755 |