siRNA Details
Gene Symbol: Olfr164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146451.1 | NP_666662.1 | 1 | 751 | ||
GenBank mRNA | BC145676.1 | - | 1 | 847 | 40130979 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146451.1 | NP_666662.1 | 1 | 751 | ||
GenBank mRNA | BC145676.1 | - | 1 | 847 | 40130979 |