siRNA Details
Gene Symbol: Olfr693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146453.2 | NP_666664.1 | 1 | 477 | ||
GenBank mRNA | BC130248.1 | - | 1 | 480 | 40142368 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr693 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146453.2 | NP_666664.1 | 1 | 477 | ||
GenBank mRNA | BC130248.1 | - | 1 | 480 | 40142368 |