siRNA Details
| Gene Symbol: Olfr722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146494.2 | NP_666705.1 | 2 | 459 | ||
| GenBank mRNA | BC051403.1 | - | 2 | 459 | 6392119 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr722| Gene Symbol: Olfr722 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146494.2 | NP_666705.1 | 2 | 459 | ||
| GenBank mRNA | BC051403.1 | - | 2 | 459 | 6392119 | |