siRNA Details
| Gene Symbol: Olfr107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146511.2 | NP_666722.2 | 1 | 578 | ||
| GenBank mRNA | BC141327.1 | - | 1 | 578 | 9055939 | |
| BC141329.1 | - | 1 | 578 | 9055941 | ||
| BC148321.1 | - | 1 | 505 | 40054936 | ||

Olfr107
Genomic Map