siRNA Details
| Gene Symbol: Olfr315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146538.2 | NP_666749.1 | 1 | 616 | ||
| GenBank mRNA | BC119148.1 | - | 1 | 616 | 8733897 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr315| Gene Symbol: Olfr315 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146538.2 | NP_666749.1 | 1 | 616 | ||
| GenBank mRNA | BC119148.1 | - | 1 | 616 | 8733897 | |