siRNA Details
Gene Symbol: Olfr315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146538.2 | NP_666749.1 | 1 | 65 | ||
GenBank mRNA | BC119148.1 | - | 1 | 65 | 8733897 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146538.2 | NP_666749.1 | 1 | 65 | ||
GenBank mRNA | BC119148.1 | - | 1 | 65 | 8733897 |