siRNA Details
| Gene Symbol: Olfr1020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146580.2 | NP_666791.1 | 1 | 203 | ||
| GenBank mRNA | BC119145.1 | - | 1 | 201 | 8733894 | |
| BC119147.1 | - | 1 | 203 | 8733896 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1020| Gene Symbol: Olfr1020 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146580.2 | NP_666791.1 | 1 | 203 | ||
| GenBank mRNA | BC119145.1 | - | 1 | 201 | 8733894 | |
| BC119147.1 | - | 1 | 203 | 8733896 | ||