siRNA Details
Gene Symbol: Olfr1046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146582.2 | NP_666793.2 | 1 | 777 | ||
GenBank mRNA | BC099891.1 | - | 1 | 776 | ||
BC099892.1 | - | 1 | 776 | |||
BC103540.1 | - | 1 | 776 |
