siRNA Details
Gene Symbol: Olfr1030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146588.2 | NP_666799.1 | 2 | 646 | ||
GenBank mRNA | BC128001.1 | - | 1 | 617 | 40135864 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr1030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146588.2 | NP_666799.1 | 2 | 646 | ||
GenBank mRNA | BC128001.1 | - | 1 | 617 | 40135864 |