siRNA Details
| Gene Symbol: Olfr433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146717.2 | NP_666928.2 | 1 | 632 | ||
| GenBank mRNA | AK154347.1 | - | 1448 | |||
| BC119238.1 | - | 1 | 632 | 8733987 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr433| Gene Symbol: Olfr433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146717.2 | NP_666928.2 | 1 | 632 | ||
| GenBank mRNA | AK154347.1 | - | 1448 | |||
| BC119238.1 | - | 1 | 632 | 8733987 | ||