siRNA Details
Gene Symbol: Olfr512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146724.1 | NP_666935.1 | 1 | 830 | ||
GenBank mRNA | BC116968.1 | - | 1 | 865 | 40126287 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146724.1 | NP_666935.1 | 1 | 830 | ||
GenBank mRNA | BC116968.1 | - | 1 | 865 | 40126287 |