siRNA Details
| Gene Symbol: Olfr512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146724.1 | NP_666935.1 | 1 | 106 | ||
| GenBank mRNA | BC116968.1 | - | 1 | 141 | 40126287 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr512| Gene Symbol: Olfr512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146724.1 | NP_666935.1 | 1 | 106 | ||
| GenBank mRNA | BC116968.1 | - | 1 | 141 | 40126287 | |