siRNA Details
| Gene Symbol: Olfr1362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146744.2 | NP_666955.2 | 1 | 514 | ||
| GenBank mRNA | BC119336.1 | - | 1 | 514 | 8734085 | |
| BC119338.1 | - | 1 | 514 | 8734087 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1362| Gene Symbol: Olfr1362 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146744.2 | NP_666955.2 | 1 | 514 | ||
| GenBank mRNA | BC119336.1 | - | 1 | 514 | 8734085 | |
| BC119338.1 | - | 1 | 514 | 8734087 | ||