siRNA Details
| Gene Symbol: Olfr551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146755.2 | NP_666966.1 | 1 | 285 | ||
| GenBank mRNA | BC141014.1 | - | 1 | 286 | 9055626 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr551| Gene Symbol: Olfr551 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146755.2 | NP_666966.1 | 1 | 285 | ||
| GenBank mRNA | BC141014.1 | - | 1 | 286 | 9055626 | |