siRNA Details
| Gene Symbol: Olfr214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146759.1 | NP_666970.1 | 1 | 242 | ||
| GenBank mRNA | BC138076.1 | - | 1 | 264 | 8861095 | |
| BC138077.1 | - | 1 | 264 | 8861096 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr214| Gene Symbol: Olfr214 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146759.1 | NP_666970.1 | 1 | 242 | ||
| GenBank mRNA | BC138076.1 | - | 1 | 264 | 8861095 | |
| BC138077.1 | - | 1 | 264 | 8861096 | ||