siRNA Details
| Gene Symbol: Olfr1176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146771.1 | NP_666982.1 | 1 | 908 | ||
| GenBank mRNA | BC127992.1 | - | 1 | 912 | 40135702 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr1176| Gene Symbol: Olfr1176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146771.1 | NP_666982.1 | 1 | 908 | ||
| GenBank mRNA | BC127992.1 | - | 1 | 912 | 40135702 | |