siRNA Details
| Gene Symbol: Olfr655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146820.2 | NP_667031.2 | 1 | 602 | ||
| GenBank mRNA | BC114416.1 | - | 1 | 601 | 40040785 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr655| Gene Symbol: Olfr655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146820.2 | NP_667031.2 | 1 | 602 | ||
| GenBank mRNA | BC114416.1 | - | 1 | 601 | 40040785 | |