siRNA Details
| Gene Symbol: Olfr640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146822.2 | NP_667033.2 | 1 | 920 | ||
| GenBank mRNA | BC106802.2 | - | 1 | 920 | 40042505 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr640| Gene Symbol: Olfr640 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146822.2 | NP_667033.2 | 1 | 920 | ||
| GenBank mRNA | BC106802.2 | - | 1 | 920 | 40042505 | |