siRNA Details
| Gene Symbol: Olfr275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146858.2 | NP_667069.2 | 1 | 944 | ||
| GenBank mRNA | BC101951.1 | - | 1 | 943 | ||
| BC141896.1 | - | 1 | 936 | |||
More Information
Other Species Targeted
Set Membership: |
|

Olfr275
Genomic Map