siRNA Details
| Gene Symbol: Olfr992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146865.1 | NP_667076.1 | 1 | 449 | ||
| GenBank mRNA | BC115959.1 | - | 1 | 448 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr992| Gene Symbol: Olfr992 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146865.1 | NP_667076.1 | 1 | 449 | ||
| GenBank mRNA | BC115959.1 | - | 1 | 448 | ||