siRNA Details
Gene Symbol: Olfr992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146865.1 | NP_667076.1 | 1 | 908 | ||
GenBank mRNA | BC115959.1 | - | 1 | 907 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr992 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146865.1 | NP_667076.1 | 1 | 908 | ||
GenBank mRNA | BC115959.1 | - | 1 | 907 |