siRNA Details
| Gene Symbol: Olfr131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146867.1 | NP_667078.1 | 1 | 458 | ||
| GenBank mRNA | BC138113.1 | - | 1 | 481 | 8861132 | |
| BC145790.1 | - | 1 | 481 | 40131093 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr131| Gene Symbol: Olfr131 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146867.1 | NP_667078.1 | 1 | 458 | ||
| GenBank mRNA | BC138113.1 | - | 1 | 481 | 8861132 | |
| BC145790.1 | - | 1 | 481 | 40131093 | ||